Learn More
Alkali Scientific L Arginine. Amino Acid, 1 Kg
SDP
Catalog No. 5000659840
Encompass_Preferred
L-Arginine, Amino Acid Grade, is an essential amino acid available in a 1kg quantity, widely used in pharmaceutical and biochemical research. L-Arginine plays a pivotal role in the synthesis of nitric oxide, which is involved in numerous physiological processes such as vasodilation and immune function. This amino acid is also utilized in protein synthesis and the production of creatine, essential for energy metabolism. In biochemical and pharmaceutical research, L-Arginine is often used in enzyme assays, synthesis of peptides, and studies related to nitric oxide production. This high-quality reagent is suitable for use in a range of biological and analytical applications.
Catalog No. 50-006-59840 Supplier Alkali Scientific Supplier No. LAR001CP
L-Arginine, Amino Acid Grade, is an essential amino acid available in a 1kg quantity, widely used in pharmaceutical and biochemical research. L-Arginine plays a pivotal role in the synthesis of nitric oxide, which is involved in numerous physiological processes such as vasodilation and immune function. This amino acid is also utilized in protein synthesis and the production of creatine, essential for energy metabolism. In biochemical and pharmaceutical research, L-Arginine is often used in enzyme assays, synthesis of peptides, and studies related to nitric oxide production. This high-quality reagent is suitable for use in a range of biological and analytical applications.