Learn More
Medchemexpress LLC L-Arginine butanoate | 80407-72-3 | 291.67 | 5 MG
SDP
Catalog No. 5000389862
Encompass_Preferred
This compound is composed of L-Arginine and butanoate. L-Arginine serves as an essential nutrient for the human body, involved in various biochemical processes. Butanoate, a short-chain fatty acid, is frequently utilized as a food additive and a solvent in pharmaceutical formulations.
  • Consists of two components: L-Arginine and butanoate.
  • L-Arginine is an essential nutrient.
  • L-Arginine participates in various biochemical processes.
  • Butanoate is a short-chain fatty acid.
  • Butanoate is commonly used as a food additive.
  • Butanoate acts as a solvent in pharmaceutical formulations.
Catalog No. 50-003-89862 Supplier Medchemexpress LLC Supplier No. HYN0455D5MG
This compound is composed of L-Arginine and butanoate. L-Arginine serves as an essential nutrient for the human body, involved in various biochemical processes. Butanoate, a short-chain fatty acid, is frequently utilized as a food additive and a solvent in pharmaceutical formulations.
  • Consists of two components: L-Arginine and butanoate.
  • L-Arginine is an essential nutrient.
  • L-Arginine participates in various biochemical processes.
  • Butanoate is a short-chain fatty acid.
  • Butanoate is commonly used as a food additive.
  • Butanoate acts as a solvent in pharmaceutical formulations.