Learn More
Medchemexpress LLC L-arginine (butanoate) | 80407-72-3 | 98.0% | 291.67 g·mol⁻¹ | C6H14N4O2 · 4/3 C4H8O2 | 10 MG
SDP
Catalog No. 5000295808
Encompass_Preferred
L-Arginine butanoate is a crystalline salt of L-arginine and butanoate supplied as a white to off-white solid for research use. It dissolves readily in water and is intended for laboratory biochemical applications requiring an arginine salt form.
  • Purity 98.0% suitable for research applications.
  • High aqueous solubility (100 mg/mL) facilitates solution preparation.
  • Solid appearance, white to off-white, for ease of handling.
  • Powder is stable under recommended storage (-20°C for long-term storage).
  • Available in small mg-scale quantities for experimental work.
Catalog No. 50-002-95808 Supplier Medchemexpress LLC Supplier No. HYN0455D10MG
L-Arginine butanoate is a crystalline salt of L-arginine and butanoate supplied as a white to off-white solid for research use. It dissolves readily in water and is intended for laboratory biochemical applications requiring an arginine salt form.
  • Purity 98.0% suitable for research applications.
  • High aqueous solubility (100 mg/mL) facilitates solution preparation.
  • Solid appearance, white to off-white, for ease of handling.
  • Powder is stable under recommended storage (-20°C for long-term storage).
  • Available in small mg-scale quantities for experimental work.