Learn More
Medchemexpress LLC L-Arginine-d7 hydrochloride | 204244-77-9 | 217.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0455AS25MG
Encompass_Preferred
This product is L-Arginine-d7 hydrochloride, a deuterium-labeled form of L-Arginine hydrochloride. L-Arginine hydrochloride serves as a nitrogen donor essential for the synthesis of nitric oxide, a powerful vasodilator often deficient in conditions like sickle cell crisis. This compound is intended for research use only.
  • Can be used as a tracer in research applications.
  • Suitable as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution may influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88807