Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Arginine from non-animal source, meets EP, USP testing specifications, suitable for cell culture | 74-79-3 | MFCD00002635 | 1kg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A80941KG
Encompass
L-Arginine from non-animal source, meets EP, USP testing specifications, suitable for cell culture | Purity: 98.5-101.0% | MW: 174.2 | 74-79-3 | MFCD00002635 | 1kg
Catalog No. 50-185-5977