Learn More
Selleck Chemical LLC L-Arginine HCl (L-Arg) S3174-100mg
SDP
Catalog No. 503109309
Encompass_Preferred
L-Arginine(L-Arg (S)-( )-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide a potent vasodilator that is deficient during times of sickle cell crisis L-Arginine HCl (L-Arg) can be used to induce animal models of Acute Pancreatitis
Catalog No. 50-310-9309 Supplier Selleck Chemical LLC Supplier No. S3174100MG
L-Arginine(L-Arg (S)-( )-Arginine hydrochloride) is the nitrogen donor for synthesis of nitric oxide a potent vasodilator that is deficient during times of sickle cell crisis L-Arginine HCl (L-Arg) can be used to induce animal models of Acute Pancreatitis