Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine HCL USP/EP/JP/FCC, Ajinomoto

Catalog No. 502946108
Change view
Click to view available options
Quantity:
1 kg
10 kg
50 kg
Packaging:
Fiber Drum
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-294-6108 50 kg Poly Drum
50-294-6032 10 kg Poly Drum
50-294-5995 1 kg Poly Drum
50-294-5976 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-294-6108 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A32602
Only null left
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 1119-34-2
Molecular Weight (g/mol) 210.66

Specifications

Packaging Poly Drum
Quantity 50 kg
Percent Purity 98.5 - 101.0%
Chemical Name or Material L-Arginine Monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.