Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, Hydrochloride, MP Biomedicals™

Catalog No. ICN10074391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10074391 1 kg
ICN10074380 100 g
ICN10074390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10074391 Supplier MP Biomedicals, Inc Supplier No. 0210074391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1119-34-2 | C6H15ClN4O2 | 210.66 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00002635
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467

Specifications

CAS 1119-34-2
Color White
Assay Percent Range 0.992
Molecular Formula C6H15ClN4O2
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
Molecular Weight (g/mol) 210.66
Quantity 1 kg
Formula Weight 210.662
Percent Purity 99.6%
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine Hydrochloride
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.