Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, Hydrochloride, MP Biomedicals™

Catalog No. MP210074396 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210074396 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210074396 Supplier MP Biomedicals, Inc Supplier No. 0210074396
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Necessary for the production of protein, helps rid the body of ammonia and stimulates insulin release. L-Arginine, Hydrochloride, MP Biomedicals is an amino acid and substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO

L-arginine is necessary for the production of protein. It also helps rid the body of ammonia (a waste product). It stimulates the release of insulin by a nitric oxide-dependent mechanism. In addition, L-arginine is used to make nitric oxide (a compound that relaxes the blood vessels).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Solubility Information Soluble in water (50mg/mL); slightly soluble in hot alcohol; insoluble in ether.
Quantity 2.5 kg
Formula Weight 210.67
Percent Purity 99.6%
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material L-Arginine Monohydrochloride
Recommended Storage Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.