Learn More
Medchemexpress LLC L-Arginine (L-glutamate) | 4320-30-3 | 321.33 | 5 G
SDP
Catalog No. 5000389072
Encompass_Preferred
L-Arginine L-glutamate is the nitrogen donor for the synthesis of nitric oxide. It can be used for research on upper gastrointestinal hypofunction or dysfunction, such as functional dyspepsia.
  • Promotes gastric emptying in rats in a dose-dependent manner.
  • Enhances adaptive relaxation of rat stomachs.
Catalog No. 50-003-89072 Supplier Medchemexpress LLC Supplier No. HYN0455B5G
L-Arginine L-glutamate is the nitrogen donor for the synthesis of nitric oxide. It can be used for research on upper gastrointestinal hypofunction or dysfunction, such as functional dyspepsia.
  • Promotes gastric emptying in rats in a dose-dependent manner.
  • Enhances adaptive relaxation of rat stomachs.