Learn More
Alkali Scientific L Arginine Monohydrochloride, 100 Grams
SDP
Catalog No. 5000660303
Encompass_Preferred
L-Arginine Monohydrochloride, 100g, is an essential amino acid and key biochemical reagent used in laboratory research, particularly in studies involving nitric oxide synthesis, protein metabolism, and cellular signaling. Arginine plays a vital role in the urea cycle, and it is also a precursor for the synthesis of creatine, proline, and other amino acids. In laboratory experiments, arginine is used to study the effects of nitric oxide in various biological systems, including immune function, vasodilation, and cell communication. The 100g package is ideal for research applications where high-quality, pure arginine is required for metabolic studies, enzyme assays, and protein synthesis research.
Catalog No. 50-006-60303 Supplier Alkali Scientific Supplier No. ARG005CP
L-Arginine Monohydrochloride, 100g, is an essential amino acid and key biochemical reagent used in laboratory research, particularly in studies involving nitric oxide synthesis, protein metabolism, and cellular signaling. Arginine plays a vital role in the urea cycle, and it is also a precursor for the synthesis of creatine, proline, and other amino acids. In laboratory experiments, arginine is used to study the effects of nitric oxide in various biological systems, including immune function, vasodilation, and cell communication. The 100g package is ideal for research applications where high-quality, pure arginine is required for metabolic studies, enzyme assays, and protein synthesis research.