Learn More
Bioworld L-Arginine, Monohydrochloride, 100 g
Catalog No. 50255284
Encompass_Preferred
A precursor of nitric oxide synthesis that induces vasodilation in vivo Chloride(%)- 16.5-17.1 Chromatographic Purity(P/F)- Pass Heavy Metals(%)- <=0.002 Identification(P/F)- Pass Loss on Drying(%)- <=0.2 Organic Volatile Impurities(P/F)- Pass Residue after Ignition(%)- <=0.1 Specific Rotation(degrees)- +21.4-+23.6 Sulfate(%)- <=0.03Appearance: White crystalline powderDensity : 1.42 g/cm3HS Code : 29252000Melting Point: 226-230°CRTECS Number: CF1995500
Catalog No. 50-255-284 Supplier Bioworld Supplier No. 401002721
A precursor of nitric oxide synthesis that induces vasodilation in vivo Chloride(%)- 16.5-17.1 Chromatographic Purity(P/F)- Pass Heavy Metals(%)- <=0.002 Identification(P/F)- Pass Loss on Drying(%)- <=0.2 Organic Volatile Impurities(P/F)- Pass Residue after Ignition(%)- <=0.1 Specific Rotation(degrees)- +21.4-+23.6 Sulfate(%)- <=0.03Appearance: White crystalline powderDensity : 1.42 g/cm3HS Code : 29252000Melting Point: 226-230°CRTECS Number: CF1995500