Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, Hydrochloride, MP Biomedicals

Catalog No. ICN10074325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10074325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10074325 Supplier MP Biomedicals, Inc Supplier No. 0210074325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity cell culture grade reagent suitable for research and manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Quantity 25 g
Formula Weight 210.7
Percent Purity 99.6%
Grade Cell Culture
Chemical Name or Material L-Arginine, Hydrochloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.