Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Isotope Laboratories L-ARGININE-N-FMOC, PBF-OH (13C6, 99%; 15N4,99%)(CONTAINS SOLVENT), 1 G OF C, 1217461-89-6

Catalog No. 501630764
Encompass

L-ARGININE-N-FMOC, PBF-OH (13C6, 99%; 15N4,99%)(CONTAINS SOLVENT), 1 G OF C, 1217461-89-6

Catalog No. 50-163-0764 Supplier Cambridge Isotope Laboratories Supplier No. CNLM8474H1
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

L-ARGININE-N-FMOC, PBF-OH (13C6, 99%; 15N4,99%)(CONTAINS SOLVENT), 1 G OF C, 1217461-89-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.