Learn More
Medchemexpress LLC L-Arginine, N2-(1-oxo-3-phenylpropyl)-L-ornithyl-L-prolyl-3-cyclohexyl-D-alanyl-L-tryptophyl- | 514814-49-4 | 99.5% | 839.04 | 1 ML
SDP
Catalog No. 5000344019
Encompass_Preferred
PMX 205 is a potent complement C5a receptor (C5aR; CD88) antagonist.
  • Recruits and activates microglia and astrocytes in vitro by activating a G protein-coupled cell-surface C5aR.
  • Shows significant results in MTT assays at 24 h, 48 h, and 72 h incubation times.
  • Orally active, selective C5aR antagonist.
  • Extends survival time, reduces end-stage motor scores, and decreases astroglial proliferation in animals.
  • Reduces fibrillar plaque load and hyperphosphorylated tau in murine models of Alzheimer's disease.
Catalog No. 50-003-44019 Supplier Medchemexpress LLC Supplier No. HY1101361MG
PMX 205 is a potent complement C5a receptor (C5aR; CD88) antagonist.
  • Recruits and activates microglia and astrocytes in vitro by activating a G protein-coupled cell-surface C5aR.
  • Shows significant results in MTT assays at 24 h, 48 h, and 72 h incubation times.
  • Orally active, selective C5aR antagonist.
  • Extends survival time, reduces end-stage motor scores, and decreases astroglial proliferation in animals.
  • Reduces fibrillar plaque load and hyperphosphorylated tau in murine models of Alzheimer's disease.