Learn More
Medchemexpress LLC L-Arginine, N2-(1-oxo-3-phenylpropyl)-L-ornithyl-L-prolyl-3-cyclohexyl-D-alanyl-L-tryptophyl- | 514814-49-4 | 99.5% | 839.04 | 25 MG
SDP
Catalog No. 5000345909
Encompass_Preferred
PMX 205 is a potent and orally active complement C5a receptor (C5aR; CD88) antagonist. This selective C5aR antagonist is supplied in solid form as a white to off-white powder.
  • Potent complement C5a receptor (C5aR; CD88) antagonist
  • Orally active and selective C5aR antagonist
  • Showed significant results in MTT assays
  • Extended survival time and reduced end-stage motor scores in a rat model of amyotrophic lateral sclerosis
  • Reduced astroglial proliferation in SOD1G93A rats
  • Reduced fibrillar plaque load in Tg2576 mice
  • Reduced hyperphosphorylated tau (69%) in 3xTg mice
  • Blocked C5a-stimulated increase in CD44 expression in PECs
  • Reversed C5a-increased PEC-derived COL4A2 secretion
  • Reduced C5a-increased proliferation of PECs
  • Induced suppression of GPX4 expression and inhibition of tumor growth in mice
Catalog No. 50-003-45909 Supplier Medchemexpress LLC Supplier No. 25MGHY110136
PMX 205 is a potent and orally active complement C5a receptor (C5aR; CD88) antagonist. This selective C5aR antagonist is supplied in solid form as a white to off-white powder.
  • Potent complement C5a receptor (C5aR; CD88) antagonist
  • Orally active and selective C5aR antagonist
  • Showed significant results in MTT assays
  • Extended survival time and reduced end-stage motor scores in a rat model of amyotrophic lateral sclerosis
  • Reduced astroglial proliferation in SOD1G93A rats
  • Reduced fibrillar plaque load in Tg2576 mice
  • Reduced hyperphosphorylated tau (69%) in 3xTg mice
  • Blocked C5a-stimulated increase in CD44 expression in PECs
  • Reversed C5a-increased PEC-derived COL4A2 secretion
  • Reduced C5a-increased proliferation of PECs
  • Induced suppression of GPX4 expression and inhibition of tumor growth in mice