Learn More
Medchemexpress LLC L-Arginine, N2-(1-oxo-3-phenylpropyl)-L-ornithyl-L-prolyl-3-cyclohexyl-D-alanyl-L-tryptophyl | 514814-49-4 | MFCD31657373 | >98.0% | 839.04 g/mol | C45H62N10O6 | 5 MG
SDP
Catalog No. 5000264199
Encompass_Preferred
PMX 205 is a cyclic peptide research compound that acts as a potent antagonist of the complement C5a receptor (C5aR; CD88). It is supplied for laboratory research use only, and is characterized by a molecular formula of C45H62N10O6 and a molecular weight of 839.04 g/mol. The product is offered in multiple pack sizes, including milligram quantities and a 10 mM solution in DMSO, and is accompanied by product datasheet and COA documentation.
Catalog No. 50-002-64199 Supplier Medchemexpress LLC Supplier No. HY1101365MG
PMX 205 is a cyclic peptide research compound that acts as a potent antagonist of the complement C5a receptor (C5aR; CD88). It is supplied for laboratory research use only, and is characterized by a molecular formula of C45H62N10O6 and a molecular weight of 839.04 g/mol. The product is offered in multiple pack sizes, including milligram quantities and a 10 mM solution in DMSO, and is accompanied by product datasheet and COA documentation.