Learn More
Medchemexpress LLC L-Arginine (Standard) | 74-79-3 | MFCD00002635 | 174.20 | 250 MG
SDP
Catalog No. 5000389934
Encompass_Preferred
L-Arginine (Standard) is the analytical standard of L-Arginine, intended for research and analytical applications. It serves as a substrate for endothelial nitric oxide synthase (eNOS) to produce NO, and is transported into vascular smooth muscle cells where it is metabolized. This compound is also a potent vasodilator and can induce experimental acute pancreatitis.
  • Analytical standard for L-Arginine
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-89934 Supplier Medchemexpress LLC Supplier No. 250MGHYN0455R
L-Arginine (Standard) is the analytical standard of L-Arginine, intended for research and analytical applications. It serves as a substrate for endothelial nitric oxide synthase (eNOS) to produce NO, and is transported into vascular smooth muscle cells where it is metabolized. This compound is also a potent vasodilator and can induce experimental acute pancreatitis.
  • Analytical standard for L-Arginine
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS