Learn More
Cayman Chemical L-Arginine (hydrochloride), Purity: ≥98%, CAS: 1119-34-2, 25 g
SDP
Catalog No. 502994532
Encompass_Preferred
An amino acid and precursor of NO; a substrate for NO synthase; enhances NO release in porcine aortic endothelial cells; induces vasodilation, increases blood flow, and reduces infarct size by 35 and 28% in normotensive and spontaneously hypertensive rats, respectively
Catalog No. 50-299-4532 Supplier Cayman Chemical Supplier No. 2370325 G
An amino acid and precursor of NO; a substrate for NO synthase; enhances NO release in porcine aortic endothelial cells; induces vasodilation, increases blood flow, and reduces infarct size by 35 and 28% in normotensive and spontaneously hypertensive rats, respectively