Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Arginyl-L-lysyl-L-histidine compound with acetic acid (1:x) | 99.5% | 439.51 (free acid) | 10 MG
SDP

Catalog No. 5000424419
Encompass_Preferred

RKH acetate is a peptide, chemically known as L-Arginyl-L-lysyl-L-histidine compound with acetic acid. It exhibits protective effects against sepsis-induced death and organ damage by directly binding to Toll-like receptor 4 (TLR4) and blocking its signal transduction within immune cells.

  • Exerts protective effects against sepsis-induced death and organ damage.
  • Directly binds to Toll-like receptor 4 (TLR4).
  • Blocks TLR4 signal transduction in immune cells.
  • Target: Toll-like receptor (TLR).
  • Pathway: Immunology/inflammation.
  • Molecular formula: C18H33N9O4.xC2H4O2.

Catalog No. 50-004-24419 Supplier Medchemexpress LLC Supplier No. HYP10208A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RKH acetate is a peptide, chemically known as L-Arginyl-L-lysyl-L-histidine compound with acetic acid. It exhibits protective effects against sepsis-induced death and organ damage by directly binding to Toll-like receptor 4 (TLR4) and blocking its signal transduction within immune cells.

  • Exerts protective effects against sepsis-induced death and organ damage.
  • Directly binds to Toll-like receptor 4 (TLR4).
  • Blocks TLR4 signal transduction in immune cells.
  • Target: Toll-like receptor (TLR).
  • Pathway: Immunology/inflammation.
  • Molecular formula: C18H33N9O4.xC2H4O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.