Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-arginylglycyl-L-aspartyl-L-serine | 91037-65-9 | 99.1% | 5 MG
SDP

Catalog No. 5000232119
Encompass_Preferred

Arg-Gly-Asp-Ser (RGDS) is a tetrapeptide integrin-binding sequence used to inhibit integrin receptor interactions and modulate cell attachment and signaling. It is supplied as a white to off-white solid with high purity and recommended cold storage for long-term stability.

  • Sequence: Arg-Gly-Asp-Ser (RGDS)
  • Cas number: 91037-65-9
  • Molecular formula: C15H27N7O8
  • Molecular weight: 433.42 g/mol
  • Purity: 99.1%
  • Appearance: white to off-white solid
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month)
  • Applications: used to study cell adhesion, integrin signaling, and fibronectin interactions

Catalog No. 50-002-32119 Supplier Medchemexpress LLC Supplier No. HY122905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arg-Gly-Asp-Ser (RGDS) is a tetrapeptide integrin-binding sequence used to inhibit integrin receptor interactions and modulate cell attachment and signaling. It is supplied as a white to off-white solid with high purity and recommended cold storage for long-term stability.

  • Sequence: Arg-Gly-Asp-Ser (RGDS)
  • Cas number: 91037-65-9
  • Molecular formula: C15H27N7O8
  • Molecular weight: 433.42 g/mol
  • Purity: 99.1%
  • Appearance: white to off-white solid
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month)
  • Applications: used to study cell adhesion, integrin signaling, and fibronectin interactions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.