Learn More
Medchemexpress LLC L-Ascorbic-6,6-C-d2 acid | 82977-10-4 | 99.90% | 5 MG
SDP
Catalog No. 5000406231
Encompass_Preferred
L-Ascorbic acid-d2 is the deuterium-labeled L-Ascorbic acid. This compound acts as an electron donor and is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and is also a collagen.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06231 Supplier Medchemexpress LLC Supplier No. HYB0166S65MG
L-Ascorbic acid-d2 is the deuterium-labeled L-Ascorbic acid. This compound acts as an electron donor and is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and is also a collagen.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS