Learn More
AdipoGen L-Ascorbic acid
SDP
Supplier:  AdipoGen CDXL0054G100
Encompass_Preferred
Chemical. CAS 50-81-7. Formula C6H8O6. MW 176.12. L-Ascorbic acid better known as Vitamin C is a naturally occurring electron donor and therefore serves as a reducing agent. It is synthesized from glucose in the liver of most mammalian species, excluding humans, non-human primates, or guinea pigs who must obtain it through dietary consumption. In humans, L-Ascorbic acid acts as an electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis which aids in the generation of adenosine triphosphate, norepinephrine synthesis, tyrosine metabolism and amidating peptides. L-Ascorbic acid demonstrates antioxidant activity that may be of some benefit for reducing the risk of developing chronic diseases such as cancer, cardiovascular disease, and cataracts. L-Ascorbic acid is an inhibitor of Cav3.2 channels, but displays no effect on Cav3.1 or Cav3.3 channels heterologously expressed in HEK 293 cells.
Catalog No. 50-256-0604