Learn More
Medchemexpress LLC L-ascorbic acid-13C-1 | 178101-89-8 | MFCD08459691 | 99.9% | 177.12 g·mol⁻¹ | C5^13CH8O6 | 10 MG
SDP
Catalog No. 5000298944
Encompass_Preferred
L-ascorbic acid-13C-1 is a 13C-labeled form of L-ascorbic acid (vitamin C) supplied as a high-purity, isotopically enriched reagent for analytical and biochemical research. It acts as an electron donor and antioxidant and is used in studies requiring isotopic labeling, including NMR and mass spectrometry.
  • 13C-labeled for isotope tracing and spectroscopic analysis.
  • High purity (99.9%) suitable for analytical and research use.
  • Available in small, research-friendly quantities (1 mg, 5 mg, 10 mg).
  • Suitable for NMR, mass spectrometry, and metabolic tracing applications.
  • Provided as a stable solid; store under recommended conditions.
Catalog No. 50-002-98944 Supplier Medchemexpress LLC Supplier No. HYB0166S210MG
L-ascorbic acid-13C-1 is a 13C-labeled form of L-ascorbic acid (vitamin C) supplied as a high-purity, isotopically enriched reagent for analytical and biochemical research. It acts as an electron donor and antioxidant and is used in studies requiring isotopic labeling, including NMR and mass spectrometry.
  • 13C-labeled for isotope tracing and spectroscopic analysis.
  • High purity (99.9%) suitable for analytical and research use.
  • Available in small, research-friendly quantities (1 mg, 5 mg, 10 mg).
  • Suitable for NMR, mass spectrometry, and metabolic tracing applications.
  • Provided as a stable solid; store under recommended conditions.