Learn More
Medchemexpress LLC L-Ascorbic acid-13C | 178101-88-7 | 99.9% | 1 MG
SDP
Catalog No. 5000406310
Encompass_Preferred
L-Ascorbic acid-13C is the 13C-labeled L-Ascorbic acid, an electron donor, and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. This compound also exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM.
  • Enhances collagen deposition and inhibits elastogenesis.
Catalog No. 50-004-06310 Supplier Medchemexpress LLC Supplier No. HYB0166S11MG
L-Ascorbic acid-13C is the 13C-labeled L-Ascorbic acid, an electron donor, and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. This compound also exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM.
  • Enhances collagen deposition and inhibits elastogenesis.