Learn More
Medchemexpress LLC L-Ascorbic acid-13C | 178101-88-7 | 99.9% | 5 MG
SDP
Catalog No. 5000406317
Encompass_Preferred
L-Ascorbic acid-13C is the 13C-labeled L-Ascorbic acid. This compound acts as an electron donor and an endogenous antioxidant agent, selectively inhibiting Cav3.2 channels. It also enhances collagen deposition, inhibits elastogenesis, and demonstrates anti-cancer effects by generating reactive oxygen species (ROS) to selectively damage cancer cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06317 Supplier Medchemexpress LLC Supplier No. HYB0166S25MG
L-Ascorbic acid-13C is the 13C-labeled L-Ascorbic acid. This compound acts as an electron donor and an endogenous antioxidant agent, selectively inhibiting Cav3.2 channels. It also enhances collagen deposition, inhibits elastogenesis, and demonstrates anti-cancer effects by generating reactive oxygen species (ROS) to selectively damage cancer cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.