Learn More
Medchemexpress LLC L-Ascorbic acid-13C-2 | 1313730-17-4 | 95.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0166S35MG
Encompass_Preferred
L-Ascorbic acid-13C-2 is the 13C labeled L-Ascorbic acid, an electron donor and endogenous antioxidant agent. It inhibits selectively Cav3.2 channels with an IC50 of 6.5 μM and is also a collagen. This compound is useful in various analytical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05251