Learn More
Medchemexpress LLC L-Ascorbic acid-13C6 | 1354064-87-1 | 99.9% | 1 MG
SDP
Catalog No. 5000406469
Encompass_Preferred
L-Ascorbic acid-13C6 is the 13C-labeled version of L-Ascorbic acid. It functions as an electron donor and is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM, acts as a collagen deposition enhancer, and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
Catalog No. 50-004-06469 Supplier Medchemexpress LLC Supplier No. HYB0166S1MG
L-Ascorbic acid-13C6 is the 13C-labeled version of L-Ascorbic acid. It functions as an electron donor and is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM, acts as a collagen deposition enhancer, and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only