Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ascorbic acid-13C6 | 1354064-87-1 | 99.9% | 182.08 | 13C6H8O6 | 10 MG
SDP

Catalog No. 5000298838
Encompass_Preferred

L-ascorbic acid-13C6 is the stable 13C6-labeled form of L-ascorbic acid supplied as a solid for use as an internal standard, tracer, or analytical standard in mass spectrometry and metabolic studies.

  • Stable 13C6 isotope labeling for tracer and internal standard applications.
  • High purity (99.9%) suitable for analytical use.
  • Formula: 13C6H8O6; molecular weight: 182.08.
  • White to light yellow solid appearance.
  • Recommended storage: powder at -20°C; in solution at -80°C for long-term storage.
  • Available as a 10 MG unit for small-scale analytical workflows.

Catalog No. 50-002-98838 Supplier Medchemexpress LLC Supplier No. HYB0166S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ascorbic acid-13C6 is the stable 13C6-labeled form of L-ascorbic acid supplied as a solid for use as an internal standard, tracer, or analytical standard in mass spectrometry and metabolic studies.

  • Stable 13C6 isotope labeling for tracer and internal standard applications.
  • High purity (99.9%) suitable for analytical use.
  • Formula: 13C6H8O6; molecular weight: 182.08.
  • White to light yellow solid appearance.
  • Recommended storage: powder at -20°C; in solution at -80°C for long-term storage.
  • Available as a 10 MG unit for small-scale analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.