Learn More
Medchemexpress LLC L-Ascorbic acid-13C6 | 1354064-87-1 | 99.9% | 5 MG
SDP
Catalog No. 5000405353
Encompass_Preferred
L-Ascorbic acid-13C6 is the 13C-labeled L-Ascorbic acid, an electron donor and endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and acts as a collagen deposition enhancer and elastogenesis inhibitor. It also exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05353 Supplier Medchemexpress LLC Supplier No. HYB0166S5MG
L-Ascorbic acid-13C6 is the 13C-labeled L-Ascorbic acid, an electron donor and endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and acts as a collagen deposition enhancer and elastogenesis inhibitor. It also exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS