Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate | 23313-12-4 | MFCD16621750 | 256.10 g/mol | C6H9O9P | 100 MG
SDP
Catalog No. 5000243283
Encompass_Preferred
L-ascorbic acid 2-phosphate is a stable, long-acting vitamin C derivative used in vitro as a cell culture supplement to stimulate collagen synthesis and support osteogenic differentiation. It increases alkaline phosphatase (ALP) activity and Runx2 expression in stem cell differentiation assays, providing a more durable source of ascorbate than L-ascorbic acid.
  • Long-acting vitamin C derivative that resists rapid oxidation.
  • Promotes collagen formation and expression.
  • Enhances osteogenic differentiation by increasing ALP activity and Runx2 expression.
  • Stable in culture media, suitable as a medium supplement.
  • Supplied as a powder for precise dosing in research applications.
Catalog No. 50-002-43283 Supplier Medchemexpress LLC Supplier No. HY103701100MG
L-ascorbic acid 2-phosphate is a stable, long-acting vitamin C derivative used in vitro as a cell culture supplement to stimulate collagen synthesis and support osteogenic differentiation. It increases alkaline phosphatase (ALP) activity and Runx2 expression in stem cell differentiation assays, providing a more durable source of ascorbate than L-ascorbic acid.
  • Long-acting vitamin C derivative that resists rapid oxidation.
  • Promotes collagen formation and expression.
  • Enhances osteogenic differentiation by increasing ALP activity and Runx2 expression.
  • Stable in culture media, suitable as a medium supplement.
  • Supplied as a powder for precise dosing in research applications.