Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ascorbic acid 2-phosphate | 23313-12-4 | MFCD16621750 | 95.0% | 256.10 g/mol | C6H9O9P | 1 G
SDP

Catalog No. 5000244194
Encompass_Preferred

L-ascorbic acid 2-phosphate (2-phospho-L-ascorbic acid) is a long-acting vitamin C derivative used in biological research and cell culture to stimulate collagen synthesis and support cellular differentiation. It is supplied as a solid reagent intended for solution preparation, assays, and tissue culture applications.

  • Long-acting vitamin C derivative that promotes collagen formation.
  • Stable solid form for easy storage and handling.
  • Soluble in aqueous solutions for cell culture and biochemical assays.
  • Molecular formula C6H9O9P, molecular weight 256.10 g/mol.
  • Available in multiple pack sizes to suit experimental needs.

Catalog No. 50-002-44194 Supplier Medchemexpress LLC Supplier No. HY1037011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ascorbic acid 2-phosphate (2-phospho-L-ascorbic acid) is a long-acting vitamin C derivative used in biological research and cell culture to stimulate collagen synthesis and support cellular differentiation. It is supplied as a solid reagent intended for solution preparation, assays, and tissue culture applications.

  • Long-acting vitamin C derivative that promotes collagen formation.
  • Stable solid form for easy storage and handling.
  • Soluble in aqueous solutions for cell culture and biochemical assays.
  • Molecular formula C6H9O9P, molecular weight 256.10 g/mol.
  • Available in multiple pack sizes to suit experimental needs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.