Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ascorbic acid 2-phosphate | 23313-12-4 | MFCD16621750 | 95.0% | 256.10 g/mol | C6H9O9P | 500 MG
SDP

Catalog No. 5000243856
Encompass_Preferred

L-ascorbic acid 2-phosphate is a stable, long-acting vitamin C derivative used as a cell culture supplement to stimulate collagen synthesis and support osteogenic differentiation. It is widely used in tissue engineering and stem cell research to enhance markers of bone formation such as alkaline phosphatase and Runx2.

  • Stable, long-acting vitamin C derivative suitable for cell culture.
  • Promotes collagen formation and extracellular matrix production.
  • Enhances osteogenic differentiation by increasing ALP activity and Runx2 expression.
  • Soluble in 1 M HCl; low solubility in water (use appropriate solvent).
  • Store at -20°C, protect from light, and store under nitrogen for stability.

Catalog No. 50-002-43856 Supplier Medchemexpress LLC Supplier No. HY103701500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ascorbic acid 2-phosphate is a stable, long-acting vitamin C derivative used as a cell culture supplement to stimulate collagen synthesis and support osteogenic differentiation. It is widely used in tissue engineering and stem cell research to enhance markers of bone formation such as alkaline phosphatase and Runx2.

  • Stable, long-acting vitamin C derivative suitable for cell culture.
  • Promotes collagen formation and extracellular matrix production.
  • Enhances osteogenic differentiation by increasing ALP activity and Runx2 expression.
  • Soluble in 1 M HCl; low solubility in water (use appropriate solvent).
  • Store at -20°C, protect from light, and store under nitrogen for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.