Learn More
Cayman Chemical L-Ascorbic Acid 2-phosphate (magnesium salt hydrate), Purity: ≥95%, CAS: 1713265-25-8, 5 g
SDP
Catalog No. 502970948
Encompass_Preferred
A long-acting ascorbic acid derivative that stimulates collagen expression and formation and is used in human cell culture; included in media to enhance the survival of human embryonic stem cells or drive osteogenic differentiation in human adipose stem cells and in human mesenchymal stromal/stem cells
Catalog No. 50-297-0948 Supplier Cayman Chemical Supplier No. 164575 G
A long-acting ascorbic acid derivative that stimulates collagen expression and formation and is used in human cell culture; included in media to enhance the survival of human embryonic stem cells or drive osteogenic differentiation in human adipose stem cells and in human mesenchymal stromal/stem cells