Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate magnesium | 113170-55-1 | MFCD11045932 | 100.0% | 292.56 | C6H9O9P · 3/2Mg | 1 G
SDP
Catalog No. 5000243230
Encompass_Preferred
L-ascorbic acid 2-phosphate magnesium is a stable, long-acting vitamin C derivative used as a cell culture supplement to promote collagen synthesis and support osteogenic differentiation. Supplied as a powdered magnesium salt, it is intended for laboratory research applications such as stem cell differentiation and tissue engineering.
  • Stable, long-acting vitamin C derivative for cell culture.
  • Stimulates collagen formation and expression.
  • Enhances osteogenic differentiation and increases alkaline phosphatase activity.
  • Supplied as a powdered magnesium salt suitable for media supplementation.
  • Available in laboratory quantities including a 1 g size for research use.
Catalog No. 50-002-43230 Supplier Medchemexpress LLC Supplier No. HY103701A1G
L-ascorbic acid 2-phosphate magnesium is a stable, long-acting vitamin C derivative used as a cell culture supplement to promote collagen synthesis and support osteogenic differentiation. Supplied as a powdered magnesium salt, it is intended for laboratory research applications such as stem cell differentiation and tissue engineering.
  • Stable, long-acting vitamin C derivative for cell culture.
  • Stimulates collagen formation and expression.
  • Enhances osteogenic differentiation and increases alkaline phosphatase activity.
  • Supplied as a powdered magnesium salt suitable for media supplementation.
  • Available in laboratory quantities including a 1 g size for research use.