Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate magnesium | 113170-55-1 | MFCD11045932 | 100.0% | 292.56 g/mol | C6H9O9P·3/2Mg | 500 MG
SDP
Catalog No. 5000244452
Encompass_Preferred
L-ascorbic acid 2-phosphate magnesium is the magnesium salt of 2-phospho-L-ascorbic acid, a stable, cell-culture-compatible derivative of vitamin C used as an antioxidant and to promote collagen synthesis. Supplied as a solid reagent for laboratory and biochemical research.
  • Stable, long-acting vitamin C derivative suitable for cell culture.
  • Promotes collagen formation and extracellular matrix production.
  • Acts as an antioxidant in biochemical assays.
  • High purity for research use (listed purity 99.97%).
  • Available in multiple pack sizes, including 500 mg.
Catalog No. 50-002-44452 Supplier Medchemexpress LLC Supplier No. HY103701A500MG
L-ascorbic acid 2-phosphate magnesium is the magnesium salt of 2-phospho-L-ascorbic acid, a stable, cell-culture-compatible derivative of vitamin C used as an antioxidant and to promote collagen synthesis. Supplied as a solid reagent for laboratory and biochemical research.
  • Stable, long-acting vitamin C derivative suitable for cell culture.
  • Promotes collagen formation and extracellular matrix production.
  • Acts as an antioxidant in biochemical assays.
  • High purity for research use (listed purity 99.97%).
  • Available in multiple pack sizes, including 500 mg.