Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate (magnesium hydrate) | 1713265-25-8 | MFCD03428049 | 99.6% | 289.5 g/mol | C6H9O9P.xH2O.3/2Mg | 100 MG
SDP
Catalog No. 5000243549
Encompass_Preferred
L-ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting vitamin C derivative used as a cell culture supplement to stimulate collagen production and support osteogenic differentiation. Supplied as a white to off-white solid with high analytical purity, it is intended for research applications requiring sustained ascorbate activity.
  • Stable, long-acting vitamin C derivative that supports collagen formation
  • Promotes alkaline phosphatase activity and osteogenic differentiation in cell culture
  • High purity (≈99.6%) suitable for research applications
  • White to off-white solid provided with certificate of analysis
  • Soluble in water (~5 mg/mL) with ultrasonic aid
  • Store sealed, away from moisture; refrigerate for long-term storage
Catalog No. 50-002-43549 Supplier Medchemexpress LLC Supplier No. HY103701B100MG
L-ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting vitamin C derivative used as a cell culture supplement to stimulate collagen production and support osteogenic differentiation. Supplied as a white to off-white solid with high analytical purity, it is intended for research applications requiring sustained ascorbate activity.
  • Stable, long-acting vitamin C derivative that supports collagen formation
  • Promotes alkaline phosphatase activity and osteogenic differentiation in cell culture
  • High purity (≈99.6%) suitable for research applications
  • White to off-white solid provided with certificate of analysis
  • Soluble in water (~5 mg/mL) with ultrasonic aid
  • Store sealed, away from moisture; refrigerate for long-term storage