Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate magnesium hydrate | 1713265-25-8 | MFCD03428049 | 99.6% | 289.5 g/mol | C6H9O9P.xH2O.3/2Mg | 500 MG
SDP
Catalog No. 5000244126
Encompass_Preferred
L-ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting derivative of ascorbic acid used primarily as a culture medium supplement and an osteogenic differentiation inducer for cell-based research. It is provided in high purity and multiple pack sizes suitable for biochemical and cell culture applications.
  • Stable, long-acting ascorbic acid derivative for sustained activity.
  • Supports osteogenic differentiation in cell culture systems.
  • Suitable as a culture medium supplement for biochemical research.
  • High purity (>99.6%) appropriate for research use.
  • Available in multiple pack sizes for flexible experimental scaling.
Catalog No. 50-002-44126 Supplier Medchemexpress LLC Supplier No. HY103701B500MG
L-ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting derivative of ascorbic acid used primarily as a culture medium supplement and an osteogenic differentiation inducer for cell-based research. It is provided in high purity and multiple pack sizes suitable for biochemical and cell culture applications.
  • Stable, long-acting ascorbic acid derivative for sustained activity.
  • Supports osteogenic differentiation in cell culture systems.
  • Suitable as a culture medium supplement for biochemical research.
  • High purity (>99.6%) appropriate for research use.
  • Available in multiple pack sizes for flexible experimental scaling.