Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate magnesium hydrate | 1713265-25-8 | MFCD03428049 | 99.6% | 579.1 g/mol | C6H9O9P·xH2O·3/2Mg | 1 G
SDP
Catalog No. 5000243284
Encompass_Preferred
L-Ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting vitamin C derivative used as a biochemical reagent and cell culture supplement. It provides an active ascorbate source with greater stability than ascorbic acid and is commonly used to promote collagen production and support osteogenic differentiation.
  • Long-acting vitamin C derivative.
  • Promotes collagen synthesis in cell culture.
  • Supports osteogenic differentiation of stem cells.
  • Greater stability than L-ascorbic acid in storage and handling.
  • High purity (~99.6%).
  • Supplied as dry powder; store sealed at 4°C, away from moisture.
  • In solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-43284 Supplier Medchemexpress LLC Supplier No. HY103701B1G
L-Ascorbic acid 2-phosphate magnesium hydrate is a stable, long-acting vitamin C derivative used as a biochemical reagent and cell culture supplement. It provides an active ascorbate source with greater stability than ascorbic acid and is commonly used to promote collagen production and support osteogenic differentiation.
  • Long-acting vitamin C derivative.
  • Promotes collagen synthesis in cell culture.
  • Supports osteogenic differentiation of stem cells.
  • Greater stability than L-ascorbic acid in storage and handling.
  • High purity (~99.6%).
  • Supplied as dry powder; store sealed at 4°C, away from moisture.
  • In solvent: -80°C (up to 6 months) or -20°C (up to 1 month).