Learn More
Apexbio Technology LLC L-Ascorbic acid 2-phosphate trisodium 66170-10-3 1g
SDP
Catalog No. 5000096290
Encompass_Preferred
L-Ascorbic acid 2-phosphate trisodium (CAS 66170-10-3) is a phosphorylated vitamin C derivative characterized by enhanced stability compared to ascorbic acid It serves as a sustained-release source of ascorbate supporting the enzymatic activity required for collagen biosynthesis Studies indicate that this compound can stimulate phosphatase activity and upregulate runx2A expression in human adipose-derived stem cells processes associated with osteogenic differentiation L-Ascorbic acid 2-phosphate trisodium is widely utilized as a medium supplement in cell culture to promote efficient osteogenesis and facilitate research into mesenchymal stem cell differentiation
Catalog No. 50-000-96290 Supplier Apexbio Technology LLC Supplier No. A46141000
L-Ascorbic acid 2-phosphate trisodium (CAS 66170-10-3) is a phosphorylated vitamin C derivative characterized by enhanced stability compared to ascorbic acid It serves as a sustained-release source of ascorbate supporting the enzymatic activity required for collagen biosynthesis Studies indicate that this compound can stimulate phosphatase activity and upregulate runx2A expression in human adipose-derived stem cells processes associated with osteogenic differentiation L-Ascorbic acid 2-phosphate trisodium is widely utilized as a medium supplement in cell culture to promote efficient osteogenesis and facilitate research into mesenchymal stem cell differentiation