Learn More
Medchemexpress LLC L-ascorbic acid 2-phosphate (trisodium) | 66170-10-3 | MFCD04038081 | >95.0% | 322.05 | C6H6Na3O9P · xH2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10783710MM 1ML
Encompass_Preferred
L-ascorbic acid 2-phosphate (trisodium) is a stable, long-acting vitamin C derivative used in cell culture and biochemical research to stimulate collagen synthesis and support extracellular matrix production. It is supplied as the trisodium salt in solid or solution formats for research use; identity corresponds to CAS 66170-10-3.
  • Stable, long-acting vitamin C derivative suitable for cell culture.
  • Promotes collagen formation and supports extracellular matrix production.
  • Trisodium salt improves solubility in aqueous solutions.
  • High purity appropriate for research applications.
  • Available in solid and solution formats for flexible use.
Catalog No. 50-002-42307