Learn More
Cayman Chemical L-AscorbIc AcId 250g
SDP
Catalog No. 502973215
Encompass_Preferred
A naturally occurring electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis (which aids in the generation of ATP), norepinephrine synthesis, tyrosine metabolism, and amidating peptides
Catalog No. 50-297-3215 Supplier Cayman Chemical Supplier No. 14656250 G
A naturally occurring electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis (which aids in the generation of ATP), norepinephrine synthesis, tyrosine metabolism, and amidating peptides