Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Ascorbic acid, 50-81-7, MFCD00064328, 25 g
Catalog No. NC1255452
Encompass
Empirical Formula (Hill Notation): C6H8O6, Molecular Weight: 176.12, meets USP testing specifications, mp: 190-194 °C, Synonym: L-Threoascorbic acid, Antiscorbutic factor, Vitamin C.
Catalog No. NC1255452 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A221825G
Empirical Formula (Hill Notation): C6H8O6, Molecular Weight: 176.12, meets USP testing specifications, mp: 190-194 °C, Synonym: L-Threoascorbic acid, Antiscorbutic factor, Vitamin C.