Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | 0044293 | 100.0% | 5 G
SDP
Catalog No. 5000406465
Encompass_Preferred
L-Ascorbic acid (L-Ascorbate), an electron donor, is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. L-Ascorbic acid also functions as a collagen deposition enhancer and an elastogenesis inhibitor. It demonstrates anti-cancer effects through the generation of reactive oxygen species (ROS) and selective damage to cancer cells.
  • Electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels (IC50 of 6.5 μM)
  • Collagen deposition enhancer
  • Elastogenesis inhibitor
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells
Catalog No. 50-004-06465 Supplier Medchemexpress LLC Supplier No. HYB01665G
L-Ascorbic acid (L-Ascorbate), an electron donor, is an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. L-Ascorbic acid also functions as a collagen deposition enhancer and an elastogenesis inhibitor. It demonstrates anti-cancer effects through the generation of reactive oxygen species (ROS) and selective damage to cancer cells.
  • Electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels (IC50 of 6.5 μM)
  • Collagen deposition enhancer
  • Elastogenesis inhibitor
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells