Learn More
Medchemexpress LLC L-ascorbic acid | 50-81-7 | 10 MG
SDP
Catalog No. 5000298220
Encompass_Preferred
GMP-grade L-ascorbic acid (vitamin C) supplied as a white to off-white solid for use as an auxiliary reagent in cell therapy manufacturing and other biochemical research. Provided in a small 10 mg package; store and handle per the certificate of analysis and safety data sheet.
  • GMP-grade material produced under good manufacturing practice guidelines.
  • White to off-white solid with formula C6H8O6.
  • Molecular weight 176.12.
  • Suitable as an auxiliary reagent for cell therapy manufacture and general research.
  • Refer to the certificate of analysis for purity and storage conditions.
Catalog No. 50-002-98220 Supplier Medchemexpress LLC Supplier No. HYB0166G10MG
GMP-grade L-ascorbic acid (vitamin C) supplied as a white to off-white solid for use as an auxiliary reagent in cell therapy manufacturing and other biochemical research. Provided in a small 10 mg package; store and handle per the certificate of analysis and safety data sheet.
  • GMP-grade material produced under good manufacturing practice guidelines.
  • White to off-white solid with formula C6H8O6.
  • Molecular weight 176.12.
  • Suitable as an auxiliary reagent for cell therapy manufacture and general research.
  • Refer to the certificate of analysis for purity and storage conditions.