Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | 50 MG
SDP
Catalog No. 5000405994
Encompass_Preferred
L-Ascorbic acid (L-Ascorbate), also known as Vitamin C, is a GMP grade small molecule produced under GMP guidelines, serving as an auxiliary reagent for cell therapy manufacture. It acts as an electron donor, an endogenous antioxidant, and an inhibitor of Cav 3.2 channels.
  • Inhibits Cav 3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS)
  • Functions as an auxiliary reagent for cell therapy manufacture
Catalog No. 50-004-05994 Supplier Medchemexpress LLC Supplier No. HYB0166G50MG
L-Ascorbic acid (L-Ascorbate), also known as Vitamin C, is a GMP grade small molecule produced under GMP guidelines, serving as an auxiliary reagent for cell therapy manufacture. It acts as an electron donor, an endogenous antioxidant, and an inhibitor of Cav 3.2 channels.
  • Inhibits Cav 3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS)
  • Functions as an auxiliary reagent for cell therapy manufacture