Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | 99.97% | 25 G
SDP
Catalog No. 5000406143
Encompass_Preferred
L-Ascorbic acid is an endogenous antioxidant agent and electron donor. It selectively inhibits Cav3.2 channels and acts as a collagen deposition enhancer and elastogenesis inhibitor. This compound demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Acts as an electron donor and endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Generates reactive oxygen species for anti-cancer effects
Catalog No. 50-004-06143 Supplier Medchemexpress LLC Supplier No. 25GHYB0166
L-Ascorbic acid is an endogenous antioxidant agent and electron donor. It selectively inhibits Cav3.2 channels and acts as a collagen deposition enhancer and elastogenesis inhibitor. This compound demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Acts as an electron donor and endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Generates reactive oxygen species for anti-cancer effects