Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | MFCD00005952 | ≥98% | 250 MG
SDP
Catalog No. 5000407410
Encompass_Preferred
L-Ascorbic acid, also known as L-Ascorbate or Vitamin C, is an electron donor and an endogenous antioxidant agent. It inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. L-Ascorbic acid exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Acts as an endogenous antioxidant agent.
  • Selectively inhibits Cav 3.2 channels with an IC50 of 6.5 μM.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Produces hypoglycaemic activity in normal and diabetic conditions when combined with Tolbutamide.
  • Action of Tolbutamide shows early onset and is maintained for a longer period in the presence of L-ascorbic acid.
Catalog No. 50-004-07410 Supplier Medchemexpress LLC Supplier No. 250MGHYB0166R
L-Ascorbic acid, also known as L-Ascorbate or Vitamin C, is an electron donor and an endogenous antioxidant agent. It inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. L-Ascorbic acid exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Acts as an endogenous antioxidant agent.
  • Selectively inhibits Cav 3.2 channels with an IC50 of 6.5 μM.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.
  • Exhibits anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Produces hypoglycaemic activity in normal and diabetic conditions when combined with Tolbutamide.
  • Action of Tolbutamide shows early onset and is maintained for a longer period in the presence of L-ascorbic acid.