Learn More
Medchemexpress LLC L-ascorbic acid | 50-81-7 | MFCD00064328 | 100.0% | 176.12 g/mol | C6H8O6 | 10 MG
SDP
Catalog No. 5000298834
Encompass_Preferred
L-ascorbic acid (vitamin C) is a water-soluble antioxidant and electron donor supplied in a GMP-like small-molecule grade for research use. It is typically provided as a white to off-white solid and is used in biochemical, cell-based, and collagen-related assays where reducing or antioxidant activity is required.
  • High purity (HPLC 99.97%).
  • GMP-like small-molecule grade.
  • White to off-white solid, water-soluble.
  • Supports antioxidant and collagen-related assays.
  • Supplied in a 10 MG unit for small-scale experiments.
Catalog No. 50-002-98834 Supplier Medchemexpress LLC Supplier No. HYB0166GL10MG
L-ascorbic acid (vitamin C) is a water-soluble antioxidant and electron donor supplied in a GMP-like small-molecule grade for research use. It is typically provided as a white to off-white solid and is used in biochemical, cell-based, and collagen-related assays where reducing or antioxidant activity is required.
  • High purity (HPLC 99.97%).
  • GMP-like small-molecule grade.
  • White to off-white solid, water-soluble.
  • Supports antioxidant and collagen-related assays.
  • Supplied in a 10 MG unit for small-scale experiments.