Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | MFCD00064328 | 99.96% | 10 G
SDP
Catalog No. 5000406087
Encompass_Preferred
L-Ascorbic acid is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels and functions as a collagen deposition enhancer and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Electron donor and endogenous antioxidant
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Generates reactive oxygen species for anti-cancer effects
  • Soluble in H2O and DMSO
  • Solid form stored at room temperature, protected from light
  • Solution stored at -80°C for 1 year or -20°C for 6 months
Catalog No. 50-004-06087 Supplier Medchemexpress LLC Supplier No. HYB016610G
L-Ascorbic acid is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels and functions as a collagen deposition enhancer and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Electron donor and endogenous antioxidant
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Generates reactive oxygen species for anti-cancer effects
  • Soluble in H2O and DMSO
  • Solid form stored at room temperature, protected from light
  • Solution stored at -80°C for 1 year or -20°C for 6 months